Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377595.2 | NP_001364843.1 | 4 - 5 | 948 | 70 | |
NR_134547.1 | 6 - 7 | 1315 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |