Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071487.1 | NP_503888.1 | 2 - 3 | 355 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |