Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071489.1 | NP_503890.1 | 5 - 6 | 660 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |