Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071683.5 | NP_504084.3 | 2 - 3 | 242 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |