Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071685.2 | NP_504086.2 | 1 - 2 | 201 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |