Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-34A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071703.4 | NP_504104.1 | 4 - 5 | 1105 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |