Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071716.2 | NP_504117.1 | 1 - 2 | 219 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |