Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322547.4 | NP_001309627.1 | 4 - 5 | 617 | 112 | |
NM_001322548.3 | NP_001309628.1 | 3 - 4 | 530 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |