Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071752.7 | NP_504153.1 | 6 - 7 | 972 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |