Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330844.1 | NP_001317803.1 | 1 - 2 | 144 | 83 | |
NM_071771.2 | NP_504172.2 | 1 - 2 | 144 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |