Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071795.1 | NP_504196.1 | 7 - 8 | 1359 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |