Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071854.4 | NP_504255.3 | 5 - 6 | 540 | 125 | |
NR_136380.1 | 7 - 8 | 827 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |