Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071867.5 | NP_504268.1 | 5 - 6 | 1486 | 82 | |
GenBank mRNA | AB010968.1 | - | 5 - 6 | 1450 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |