Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071894.4 | NP_504295.1 | 2 - 3 | 230 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |