Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071905.1 | NP_504306.1 | 2 - 3 | 555 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |