Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269125.2 | NP_001256054.1 | 5 - 6 | 878 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |