Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129504.4 | NP_001122976.1 | 5 - 6 | 777 | 64 | |
NM_071946.9 | NP_504347.2 | 6 - 7 | 846 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |