Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072014.7 | NP_504415.2 | 10 - 11 | 1887 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |