Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028472.5 | NP_001023643.1 | 5 - 6 | 1048 | 102 | |
NM_072014.7 | NP_504415.2 | 5 - 6 | 1061 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |