Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072068.6 | NP_504469.1 | 2 - 3 | 240 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |