Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hex-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072088.7 | NP_504489.3 | 8 - 9 | 1443 | 72 | |
GenBank mRNA | AM748821.2 | - | 8 - 9 | 1414 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |