Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129376.4 | NP_001122848.1 | 10 - 11 | 1808 | 69 | |
NM_072093.7 | NP_504494.2 | 9 - 10 | 1517 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |