Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyn-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129598.3 | NP_001123070.1 | 7 - 8 | 1167 | 62 | |
NM_001129599.4 | NP_001123071.1 | 7 - 8 | 1197 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |