Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072135.1 | NP_504536.1 | 4 - 5 | 789 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |