Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047644.2 | NP_001041109.1 | 4 - 5 | 448 | 134 | |
NR_136375.1 | 7 - 8 | 1051 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |