Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047644.2 | NP_001041109.1 | 6 - 7 | 638 | 140 | |
NR_136375.1 | 9 - 10 | 1241 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |