Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jamp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373030.1 | NP_001360559.1 | 1 - 2 | 324 | 114 | |
NM_072167.8 | NP_504568.1 | 1 - 2 | 324 | 114 | ||
GenBank mRNA | AF304119.1 | - | 1 - 2 | 324 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |