Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001359765.3 | NP_001346723.1 | 35375 | 97 | ||
NM_001359766.3 | NP_001346716.1 | 35375 | 97 | |||
NM_001359767.3 | NP_001346722.1 | 36653 | 97 | |||
NM_001359768.3 | NP_001346715.1 | 36653 | 97 | |||
NM_001373034.1 | NP_001360072.1 | 36746 | 97 | |||
NM_001380654.1 | NP_001368672.1 | 8 - 9 | 14663 | 97 | ||
NM_001380655.1 | NP_001368671.1 | 8 - 9 | 14663 | 97 | ||
NM_001380656.1 | NP_001367914.1 | 35375 | 97 | |||
NM_001380657.1 | NP_001367912.1 | 35468 | 97 | |||
NM_001380658.2 | NP_001367913.1 | 36653 | 97 | |||
NM_001380659.1 | NP_001368674.1 | 8 - 9 | 14663 | 97 | ||
NM_001380660.1 | NP_001368673.1 | 8 - 9 | 14663 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |