Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrck-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313212.2 | NP_001300141.1 | 2 - 3 | 289 | 69 | |
NM_072198.7 | NP_504599.2 | 2 - 3 | 199 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |