Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrck-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313212.2 | NP_001300141.1 | 9 - 10 | 4492 | 87 | |
NM_001313213.3 | NP_001300142.1 | 5 - 6 | 3556 | 87 | ||
NM_001313214.3 | NP_001300143.1 | 5 - 6 | 3562 | 87 | ||
NM_001313215.3 | NP_001300144.1 | 3 - 4 | 2365 | 87 | ||
NM_072198.7 | NP_504599.2 | 9 - 10 | 4408 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |