Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028766.4 | NP_001023937.1 | 7 - 8 | 790 | 94 | |
NM_001028767.7 | NP_001023938.1 | 6 - 7 | 688 | 94 | ||
NM_001383322.1 | 6 - 7 | 737 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |