Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028766.4 | NP_001023937.1 | 9 - 10 | 1838 | 91 | |
NM_001028767.7 | NP_001023938.1 | 8 - 9 | 1736 | 91 | ||
NM_001383322.1 | 8 - 9 | 1785 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |