Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072282.5 | NP_504683.1 | 2 - 3 | 305 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |