Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028960.5 | NP_001024131.1 | 11 - 12 | 1583 | 77 | |
NM_072288.6 | NP_504689.1 | 10 - 11 | 1446 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |