Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spn-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072316.6 | NP_504717.1 | 1 - 2 | 63 | 73 | |
GenBank mRNA | AB052819.1 | - | 1 - 2 | 66 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |