Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028882.5 | NP_001024053.2 | 10 - 11 | 6530 | 89 | |
NM_001307743.4 | NP_001294672.1 | 8 - 9 | 6427 | 89 | ||
GenBank mRNA | AF166169.1 | - | 9 - 10 | 6478 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |