Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072357.4 | NP_504758.1 | 3 - 4 | 709 | 89 |
Gene Symbol: T07H8.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368454.3 | NP_001355523.1 | 2 - 3 | 1006 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |