Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356689.2 | NP_001343632.1 | 17 - 18 | 4509 | 107 | |
NM_001383329.1 | 16 - 17 | 5401 | 107 | |||
NM_072382.9 | NP_504783.2 | 18 - 19 | 5198 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |