Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373048.3 | NP_001360011.1 | 4 - 5 | 1190 | 89 | |
NM_072394.7 | NP_504795.1 | 4 - 5 | 1229 | 89 | ||
GenBank mRNA | AY519856.1 | - | 4 - 5 | 1171 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |