Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373048.3 | NP_001360011.1 | 6 - 7 | 1937 | 85 | |
NM_072394.7 | NP_504795.1 | 6 - 7 | 1976 | 85 | ||
GenBank mRNA | AY519856.1 | - | 6 - 7 | 1918 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |