Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fat-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072413.7 | NP_504814.1 | 3 - 4 | 589 | 136 | |
GenBank mRNA | AF260243.1 | - | 3 - 4 | 574 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |