Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072419.5 | NP_504820.2 | 8 - 9 | 1409 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |