Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072483.6 | NP_504884.1 | 3 - 4 | 1201 | 80 | |
NR_131691.1 | 2 - 3 | 1150 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |