Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072484.5 | NP_504885.2 | 2 - 3 | 213 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |