Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072496.1 | NP_504897.1 | 1 - 2 | 152 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |