Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srxa-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072501.2 | NP_504902.2 | 2 - 3 | 416 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |