Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072504.1 | NP_504905.1 | 5 - 6 | 930 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |