Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072525.5 | NP_504926.2 | 1 - 2 | 135 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |