Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028564.5 | NP_001023735.1 | 7 - 8 | 937 | 94 | |
NM_001028565.4 | NP_001023736.1 | 6 - 7 | 1024 | 94 | ||
GenBank mRNA | AF497825.1 | - | 6 - 7 | 907 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |