Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269165.1 | NP_001256094.1 | 4 - 5 | 714 | 144 | |
NM_001373058.1 | NP_001360010.1 | 5 - 6 | 669 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |